Learn More
Medchemexpress LLC PRX-07034 hydrochloride | 903580-39-2 | 98.97% | C21H29Cl2N3O4S | 100 MG
SDP
Catalog No. 5000355270
Encompass_Preferred
PRX-07034 hydrochloride is a potent and highly selective 5-HT6 receptor antagonist, exhibiting a Ki of 4-8 nM and an IC50 of 19 nM. This compound is valuable for research focused on enhancing working memory and cognitive flexibility.
  • Highly selective 5-HT6 receptor antagonist.
  • Demonstrates Ki of 4-8 nM and IC50 of 19 nM for 5-HT6 receptor.
  • Supports research in enhancing working memory and cognitive flexibility.
  • Shows ≥100-fold selectivity for 5-HT6 receptor over 68 other GPCRs, ion channels, and transporters.
  • Inhibits multiple 5-HT and dopamine receptors at varying concentrations.
  • Enhances delayed spontaneous alternation and strategy switching in animal models.
Catalog No. 50-003-55270 Supplier Medchemexpress LLC Supplier No. HY14559100MG
PRX-07034 hydrochloride is a potent and highly selective 5-HT6 receptor antagonist, exhibiting a Ki of 4-8 nM and an IC50 of 19 nM. This compound is valuable for research focused on enhancing working memory and cognitive flexibility.
  • Highly selective 5-HT6 receptor antagonist.
  • Demonstrates Ki of 4-8 nM and IC50 of 19 nM for 5-HT6 receptor.
  • Supports research in enhancing working memory and cognitive flexibility.
  • Shows ≥100-fold selectivity for 5-HT6 receptor over 68 other GPCRs, ion channels, and transporters.
  • Inhibits multiple 5-HT and dopamine receptors at varying concentrations.
  • Enhances delayed spontaneous alternation and strategy switching in animal models.