Learn More
Medchemexpress LLC Prx-07034 hydrochloride | 903580-39-2 | 99.0% | 490.44 | C21H29Cl2N3O4S | 5 MG
SDP
Catalog No. 5000221288
Encompass_Preferred
PRX-07034 hydrochloride is a research-use small molecule that acts as a potent, selective antagonist of the serotonin 5-HT6 receptor. Supplied as the hydrochloride salt, it is intended for preclinical pharmacology studies investigating cognition, working memory, and receptor pharmacology. The compound is offered in multiple pack sizes and formats and is reported with high purity.
  • Potent, selective 5-HT6 receptor antagonist (Ki 4-8 nM; IC50 19 nM).
  • High reported purity (98.97%).
  • Available in multiple pack sizes and as a 10 mM solution in DMSO.
  • Suitable for preclinical studies of cognition and memory.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-21288 Supplier Medchemexpress LLC Supplier No. HY145595MG
PRX-07034 hydrochloride is a research-use small molecule that acts as a potent, selective antagonist of the serotonin 5-HT6 receptor. Supplied as the hydrochloride salt, it is intended for preclinical pharmacology studies investigating cognition, working memory, and receptor pharmacology. The compound is offered in multiple pack sizes and formats and is reported with high purity.
  • Potent, selective 5-HT6 receptor antagonist (Ki 4-8 nM; IC50 19 nM).
  • High reported purity (98.97%).
  • Available in multiple pack sizes and as a 10 mM solution in DMSO.
  • Suitable for preclinical studies of cognition and memory.
  • For research use only; not for human or veterinary use.