Learn More
Medchemexpress LLC PRX-07034 hydrochloride | 903580-39-2 | 99.0% | C21H29Cl2N3O4S | 10MG
SDP
Catalog No. 5000261635
Encompass_Preferred
PRX-07034 hydrochloride is a research-grade, selective 5-HT6 receptor antagonist supplied as a white to off-white solid. It is intended for laboratory research to support studies of cognition and memory and is not for human use.
  • Potent 5-HT6 receptor antagonist (Ki = 4-8 nM; IC50 = 19 nM).
  • High purity (98.97%) suitable for research applications.
  • White to off-white solid, easy to weigh and handle.
  • Stable when stored sealed and away from moisture; recommended storage at -20°C.
  • Intended for preclinical and in vitro research use only.
Catalog No. 50-002-61635 Supplier Medchemexpress LLC Supplier No. HY1455910MG
PRX-07034 hydrochloride is a research-grade, selective 5-HT6 receptor antagonist supplied as a white to off-white solid. It is intended for laboratory research to support studies of cognition and memory and is not for human use.
  • Potent 5-HT6 receptor antagonist (Ki = 4-8 nM; IC50 = 19 nM).
  • High purity (98.97%) suitable for research applications.
  • White to off-white solid, easy to weigh and handle.
  • Stable when stored sealed and away from moisture; recommended storage at -20°C.
  • Intended for preclinical and in vitro research use only.