Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prx-07034 hydrochloride | 903580-39-2 | 99.0% | C21H29Cl2N3O4S | 25 MG
SDP

Catalog No. 5000353118
Encompass_Preferred

PRX-07034 hydrochloride is a highly selective and potent 5-HT6 receptor antagonist with a Ki of 4-8 nM and IC50 of 19 nM. Intended for research on enhancing working memory and cognitive flexibility, it shows significant 5-HT6 receptor selectivity (>100-fold over 68 other GPCRs, ion channels, and transporters). It also inhibits 5-HT1A, 5-HT1B, 5-HT1D, Dopamine D3, Histamine H2, and Opioid μ receptors. In vivo studies in rats demonstrated enhanced delayed spontaneous alternation and improved cognitive strategy switching.

  • Highly selective and potent 5-HT6 receptor antagonist
  • Ki of 4-8 nM and IC50 of 19 nM for 5-HT6 receptor
  • Enhances working memory and cognitive flexibility
  • >100-fold selectivity over 68 other GPCRs, ion channels, and transporters
  • Inhibits 5-HT1A, 5-HT1B, 5-HT1D, Dopamine D3, Histamine H2, and Opioid μ receptors

Catalog No. 50-003-53118 Supplier Medchemexpress LLC Supplier No. 25MGHY14559
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PRX-07034 hydrochloride is a highly selective and potent 5-HT6 receptor antagonist with a Ki of 4-8 nM and IC50 of 19 nM. Intended for research on enhancing working memory and cognitive flexibility, it shows significant 5-HT6 receptor selectivity (>100-fold over 68 other GPCRs, ion channels, and transporters). It also inhibits 5-HT1A, 5-HT1B, 5-HT1D, Dopamine D3, Histamine H2, and Opioid μ receptors. In vivo studies in rats demonstrated enhanced delayed spontaneous alternation and improved cognitive strategy switching.

  • Highly selective and potent 5-HT6 receptor antagonist
  • Ki of 4-8 nM and IC50 of 19 nM for 5-HT6 receptor
  • Enhances working memory and cognitive flexibility
  • >100-fold selectivity over 68 other GPCRs, ion channels, and transporters
  • Inhibits 5-HT1A, 5-HT1B, 5-HT1D, Dopamine D3, Histamine H2, and Opioid μ receptors

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.