Learn More
Medchemexpress LLC PRX-08066 | 866206-54-4 | C19H17ClFN5S | 1 MG
SDP
Catalog No. 5000376588
Encompass_Preferred
PRX-08066 is a selective and orally active 5-hydroxytryptamine receptor 2B (5-HT2BR) antagonist. It inhibits the MAPK pathway, 5-HT release, and the expression of fibrotic factors. This product can be used for research related to pulmonary Arterial Hypertension (PAH) and neuroendocrine tumors (NET).
  • Selective and orally active 5-HT2BR antagonist (Ki of 3.4 nM)
  • Inhibits MAPK pathway and 5-HT release
  • Inhibits expression of fibrotic factors (TGFβ1, CTGF, and FGF2)
  • Inhibits proliferation of KRJ-I cells
  • Induces apoptosis via caspase-3 activation
  • Inhibits pulmonary vascular remodeling
Catalog No. 50-003-76588 Supplier Medchemexpress LLC Supplier No. HY154721MG
PRX-08066 is a selective and orally active 5-hydroxytryptamine receptor 2B (5-HT2BR) antagonist. It inhibits the MAPK pathway, 5-HT release, and the expression of fibrotic factors. This product can be used for research related to pulmonary Arterial Hypertension (PAH) and neuroendocrine tumors (NET).
  • Selective and orally active 5-HT2BR antagonist (Ki of 3.4 nM)
  • Inhibits MAPK pathway and 5-HT release
  • Inhibits expression of fibrotic factors (TGFβ1, CTGF, and FGF2)
  • Inhibits proliferation of KRJ-I cells
  • Induces apoptosis via caspase-3 activation
  • Inhibits pulmonary vascular remodeling