Learn More
Selleck Chemical LLC PRX-08066 Maleic acid
SDP
Catalog No. 5000002834
Encompass_Preferred
PRX-08066 Maleic acid is a selective 5-HT2B receptor antagonist with IC50 of 3 4 nM prevents the severity of pulmonary arterial hypertension in the MCT rat model Phase 2
Catalog No. 50-000-02834 Supplier Selleck Chemical LLC Supplier No. S80105MG
PRX-08066 Maleic acid is a selective 5-HT2B receptor antagonist with IC50 of 3 4 nM prevents the severity of pulmonary arterial hypertension in the MCT rat model Phase 2