Learn More
TARGETMOL CHEMICALS INC PRX933 HYDROCHLORIDE 5MG
SDP
Catalog No. 502581440
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PRX933 hydrochloride (GW876167 hydrochloride) is an agonist of 5-HT2C receptor and can be used in studies about hypertension acute treatment. purity: 99%
Catalog No. 50-258-1440 Supplier TARGETMOL CHEMICALS INC Supplier No. T125655MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PRX933 hydrochloride (GW876167 hydrochloride) is an agonist of 5-HT2C receptor and can be used in studies about hypertension acute treatment. purity: 99%