Learn More
Associates of Cape Cod Inc PRYOTELL LAL 0.06 ML 5ML 5PK
SDP
Catalog No. NC0243657
Encompass
PYROTELL GEL CLOT LAL, 0.06 EU/mL SENSITIVITY, 5mL/VIAL, 5 VIALS/PACK
Catalog No. NC0243657 Supplier Associates of Cape Cod Inc Supplier No. G50065
PYROTELL GEL CLOT LAL, 0.06 EU/mL SENSITIVITY, 5mL/VIAL, 5 VIALS/PACK