Learn More
Medchemexpress LLC HY-N5057 1mg Medchemexpress, Przewalskinic acid A CAS:136112-75-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN50571MG
Encompass_Preferred
Medchemexpress, HY-N5057 1mg Przewalskinic acid A CAS:136112-75-9 Przewalskinic acid A is a rare, water-soluble phenolic acid thus far only found in the Salvia przewalskii Maxim herb. Phenolic acids show potent antioxidant activities and potential effects in protecting against brain and heart damage caused by ischemia reperfusion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0441