Learn More
TARGETMOL CHEMICALS INC PS-1145 10MG
SDP
Catalog No. 502879467
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. PS-1145 (IKK Inhibitor X) is a specific IKK inhibitor with IC50 of 88 nM. Purity 99.63%
Catalog No. 50-287-9467 Supplier TARGETMOL CHEMICALS INC Supplier No. T309410MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. PS-1145 (IKK Inhibitor X) is a specific IKK inhibitor with IC50 of 88 nM. Purity 99.63%