Learn More
Apexbio Technology LLC PS-1145 431898-65-6 25mg
SDP
Catalog No. 5000088376
Encompass_Preferred
PS-1145 (CAS 431898-65-6) is a selective inhibitor of I B kinase (IKK) with an IC50 of 88 nM IKK is a critical component of the upstream NF- B signaling pathway regulating cellular responses to inflammation In vitro studies have demonstrated that PS-1145 blocks TNF -induced NF- B activation in multiple myeloma (MM) cells in a dose- and time-dependent manner increases I B protein levels suppresses IL-6-mediated anti-apoptotic effects and inhibits expression of ICAM-1 Additionally PS-1145 reduces IL-6 secretion from bone marrow stromal cells (BMSCs) and partially limits MM cell proliferation In vivo PS-1145 has been shown to induce apoptosis in DMBA-induced skin tumors in rats by inhibiting NF- B and VEGF activity The compound remains in the preclinical research stage
Catalog No. 50-000-88376 Supplier Apexbio Technology LLC Supplier No. B608925
PS-1145 (CAS 431898-65-6) is a selective inhibitor of I B kinase (IKK) with an IC50 of 88 nM IKK is a critical component of the upstream NF- B signaling pathway regulating cellular responses to inflammation In vitro studies have demonstrated that PS-1145 blocks TNF -induced NF- B activation in multiple myeloma (MM) cells in a dose- and time-dependent manner increases I B protein levels suppresses IL-6-mediated anti-apoptotic effects and inhibits expression of ICAM-1 Additionally PS-1145 reduces IL-6 secretion from bone marrow stromal cells (BMSCs) and partially limits MM cell proliferation In vivo PS-1145 has been shown to induce apoptosis in DMBA-induced skin tumors in rats by inhibiting NF- B and VEGF activity The compound remains in the preclinical research stage