Learn More
Medchemexpress LLC PS-1145 | 431898-65-6 | 99.67% | 322.75 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY18008100MG
Encompass_Preferred
PS-1145 is an IκB kinase (IKK) inhibitor with an IC50 of 88 nM, functioning to block TNFα-induced NF-κB activation by inhibiting IκBα phosphorylation in multiple myeloma (MM) cells. This inhibition is observed in a dose- and time-dependent manner. It also enhances the blockade of NF-κB activation when combined with Dexamethasone, and can inhibit the protective effect of IL-6 against Dexamethasone-induced apoptosis.
  • Potent inhibitor of IKK activity.
  • Blocks phosphorylation of IκBα.
  • Inhibits TNFα-induced NF-κB activation.
  • Reduces TNFα-induced intracellular adhesion molecule (ICAM)-1 expression.
  • Demonstrates antiproliferative activity against certain human cells.
  • Decreases serum levels of specific cytokines in vivo.
  • Can block inflammation-induced weight gain during high-fat feeding.
Catalog No. 50-003-87488