Learn More
Medchemexpress LLC PS-1145 | 431898-65-6 | 99.67% | 322.75 | 25 MG
SDP
Catalog No. 5000383901
Encompass_Preferred
PS-1145 is an IκB kinase (IKK) inhibitor with an IC50 of 88 nM.
  • Blocks TNFα-induced NF-κB activation in a dose- and time-dependent manner in MM cells through inhibition of IκBα phosphorylation.
  • Dexamethasone enhances the blockade of NF-κB activation by the product.
  • Blocks the protective effect of IL-6 against Dexamethasone-induced apoptosis.
  • Inhibits TNFα-induced intracellular adhesion molecule (ICAM)-1 expression.
  • Inhibits both IL-6 secretion from bone marrow stromal cells (BMSCs) triggered by MM cell adhesion and proliferation of MM cells adherent to BMSCs.
  • Administration results in a significant decrease in serum levels of three cytokines.
  • Blocks hypothalamic inflammation induced by IL-4.
  • Decreases IL-1β mRNA content.
Catalog No. 50-003-83901 Supplier Medchemexpress LLC Supplier No. 25MGHY18008
PS-1145 is an IκB kinase (IKK) inhibitor with an IC50 of 88 nM.
  • Blocks TNFα-induced NF-κB activation in a dose- and time-dependent manner in MM cells through inhibition of IκBα phosphorylation.
  • Dexamethasone enhances the blockade of NF-κB activation by the product.
  • Blocks the protective effect of IL-6 against Dexamethasone-induced apoptosis.
  • Inhibits TNFα-induced intracellular adhesion molecule (ICAM)-1 expression.
  • Inhibits both IL-6 secretion from bone marrow stromal cells (BMSCs) triggered by MM cell adhesion and proliferation of MM cells adherent to BMSCs.
  • Administration results in a significant decrease in serum levels of three cytokines.
  • Blocks hypothalamic inflammation induced by IL-4.
  • Decreases IL-1β mRNA content.