Learn More
Medchemexpress LLC Ps-1145 | 431898-65-6 | MFCD08705411 | 99.9% | 322.75 | C17H11ClN4O | 10 MG
SDP
Catalog No. 5000295245
Encompass_Preferred
PS-1145 is a small-molecule inhibitor of IκB kinase (IKK) used in research to block NF-κB signaling and induce apoptosis in cell models. It is supplied as a powder with documented solubility and storage recommendations for experimental use.
  • Potent IKK inhibitor with IC50 of 88 nM.
  • High chemical purity (99.88%).
  • Provided as a powder for convenient handling and formulation.
  • Soluble in DMSO at 20.83 mg/mL; multiple in vitro formulations available.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for in vitro studies of NF-κB signaling and apoptosis pathways.
Catalog No. 50-002-95245 Supplier Medchemexpress LLC Supplier No. HY1800810MG
PS-1145 is a small-molecule inhibitor of IκB kinase (IKK) used in research to block NF-κB signaling and induce apoptosis in cell models. It is supplied as a powder with documented solubility and storage recommendations for experimental use.
  • Potent IKK inhibitor with IC50 of 88 nM.
  • High chemical purity (99.88%).
  • Provided as a powder for convenient handling and formulation.
  • Soluble in DMSO at 20.83 mg/mL; multiple in vitro formulations available.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for in vitro studies of NF-κB signaling and apoptosis pathways.