Learn More
Medchemexpress LLC HY-18008 5mg Medchemexpress, PS-1145 CAS:431898-65-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY180085MG
Encompass_Preferred
Medchemexpress, HY-18008 5mg PS-1145 CAS:431898-65-6 PS-1145 is an IκB kinase (IKK) inhibitor with an IC50 of 88 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2009