Learn More
Selleck Chemical LLC PS-1145
SDP
Catalog No. 5000000404
Encompass_Preferred
PS-1145 is a specific IKK inhibitor with IC50 of 88 nM
Catalog No. 50-000-00404 Supplier Selleck Chemical LLC Supplier No. S769125MG
PS-1145 is a specific IKK inhibitor with IC50 of 88 nM