Learn More
Rotronic Instrument Corp PS-14 - 14mm (depth) Sample Cups, 100/pk
SDP
Catalog No. NC1817526 Shop All Rotronic Instrument Corp Products
Encompass_Preferred
The disposable sample containers ensure the optimum sample volume is introduced into the WP-14-S sample holders. They prevent the sample holders from coming into direct contact with the product being tested, thereby preventing soiling or cross contamination. The sample containers also provide a convenient means of collecting and storing samples.
Catalog No. NC1817526 Supplier Rotronic Instrument Corp Supplier No. 5030.PS14.01
The disposable sample containers ensure the optimum sample volume is introduced into the WP-14-S sample holders. They prevent the sample holders from coming into direct contact with the product being tested, thereby preventing soiling or cross contamination. The sample containers also provide a convenient means of collecting and storing samples.