Learn More
Electron Microscopy Sciences PS 150 XO Calibration Slide
SDP
Catalog No. 501902450
Encompass_Preferred
All of the PS-XO series of Calibration Slides are made with Green Float Glass and have a bright Chrome Image High definition 150mm scale in 0.1mm divisions with extended 0.5mm, 1.0mm and 5.0mm lines. Series of dots and crosses, at 10mm intervals.
Catalog No. 50-190-2450 Supplier Electron Microscopy Sciences Supplier No. 68073PS150
All of the PS-XO series of Calibration Slides are made with Green Float Glass and have a bright Chrome Image High definition 150mm scale in 0.1mm divisions with extended 0.5mm, 1.0mm and 5.0mm lines. Series of dots and crosses, at 10mm intervals.