Learn More
AdipoGen Bortezomib, 179324-69-7, MFCD09056737, 25 mg
SDP
Catalog No. 501016264
Encompass_Preferred
MF: C19H25BN4O4, MW: 384.2, ≥97%, Synonyms: PS-341; LDP-341; MLM-341; MG-341; NSC-681239; Proteasome Inhibitor. Appearance: White solid. Highly potent, selective and reversible cell permeable inhibitor of the proteasome. Inhibits the chymotrypsin-like and caspase-like peptidase activity of the proteasome. Calpain and cathepsin inhibitor. Autophagy activator. Anticancer compound. Inhibits proliferation and migration of several tumor cell lines with nanomolar potency. Apoptosis inducer.
Catalog No. 50-101-6264 Supplier AdipoGen Supplier No. AGCR13602M025
MF: C19H25BN4O4, MW: 384.2, ≥97%, Synonyms: PS-341; LDP-341; MLM-341; MG-341; NSC-681239; Proteasome Inhibitor. Appearance: White solid. Highly potent, selective and reversible cell permeable inhibitor of the proteasome. Inhibits the chymotrypsin-like and caspase-like peptidase activity of the proteasome. Calpain and cathepsin inhibitor. Autophagy activator. Anticancer compound. Inhibits proliferation and migration of several tumor cell lines with nanomolar potency. Apoptosis inducer.