Learn More
Ubpbio PS-341 (BORTEZOMIB), 5MG
Catalog No. 501126894
Encompass_Preferred
Inhibitors – PS-341 (UBPBio catalog# F1200, F1201, F1202) – PS-341 (also called Bortezomib) is a specific, reversible and cell permeable dipeptide boronic acid inhibitor of the chymotrypsin-like activity of the proteasome (Ki=0.6nM). It inhibits proliferation of various tumor cell lines with nanomolar potency. Its commercial version was approved by the USFDA for the treatment of relapsed multiple myeloma. The typical concentrations for cell culture use are 1-20 µM. For in vitro use, the typical concentrations are 10-100 µM. CAS# 179324-69-7.
Catalog No. 50-112-6894 Supplier Ubpbio Supplier No. F1200
Add to Cart
Edge
Add to Cart
Inhibitors – PS-341 (UBPBio catalog# F1200, F1201, F1202) – PS-341 (also called Bortezomib) is a specific, reversible and cell permeable dipeptide boronic acid inhibitor of the chymotrypsin-like activity of the proteasome (Ki=0.6nM). It inhibits proliferation of various tumor cell lines with nanomolar potency. Its commercial version was approved by the USFDA for the treatment of relapsed multiple myeloma. The typical concentrations for cell culture use are 1-20 µM. For in vitro use, the typical concentrations are 10-100 µM. CAS# 179324-69-7.