Learn More
Selleck Chemical LLC PS-48
SDP
Supplier:  Selleck Chemical LLC S758625MG
Encompass_Preferred
PS-48 is an allosteric activator of phosphoinositide-dependent protein kinase-1 (PDK1)
Catalog No. 50-000-04210