Learn More
Selleck Chemical LLC PS-48
SDP
Catalog No. 5000004211
Encompass_Preferred
PS-48 is an allosteric activator of phosphoinositide-dependent protein kinase-1 (PDK1)
Catalog No. 50-000-04211 Supplier Selleck Chemical LLC Supplier No. S75865MG
PS-48 is an allosteric activator of phosphoinositide-dependent protein kinase-1 (PDK1)