Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ps-750-M

Catalog No. 9111511G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-115-11G 1 g
91-115-125G 25 g
91-115-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 91-115-11G Supplier Sigma Aldrich Supplier No. 9111511G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced to increase the catalytic efficiency and acts as an environmentally benign and sustainable amphiphile. Find details here.PS-750-M is a custom surfactant developed in the Handa lab. PS-750-M allows for a variety of reactions, including challenging cross-couplings and monofluorination of indoles, to be conducted in water.Micelle-Enabled Palladium Catalysis for Convenient sp2-sp3 Coupling of Nitroalkanes with Aryl Bromides in Water Under Mild ConditionsShielding Effect of Micelle for Highly Effective and Selective Monofluorination of Indoles in Water

Specifications

CAS 135447-74-2
Linear Formula (C2H4O)nC18H33NO3
Quantity 1 g
Synonym FI-750-M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.