Learn More
FUJIFILM Biosciences PS Capture™ Exosome Flow Cytometry Kit, 300 tests
Catalog No. NC1578363
Encompass_Preferred
Kit Components (300 assays): Exosome Capture Beads 1 x 3 mL, Washing Buffer (10×) 2 x 45 mL, Exosome Binding Enhancer (100×) 1 x 15 mL. This kit is a reagent that can qualitatively analyze extracellular vesicles contained in cell culture supernatants and body fluid samples by flow cytometry. After extracellular vesicles are reacted and immobilized on magnetic beads on which extracellular vesicle surface phosphatidylserine (PS) specifically binding protein is immobilized, fluorescent labeling for any extracellular vesicle marker protein by using antibodies, we can detect marker proteins on extracellular vesicle surface with high sensitivity. Before using it, prepare the primary antibody for each surface marker protein, the fluorescent dye-labeled secondary antibody, or the primary antibody labeled with fluorescent dye.
Catalog No. NC1578363 Supplier FUJIFILM Biosciences Supplier No. 29779701
Kit Components (300 assays): Exosome Capture Beads 1 x 3 mL, Washing Buffer (10×) 2 x 45 mL, Exosome Binding Enhancer (100×) 1 x 15 mL. This kit is a reagent that can qualitatively analyze extracellular vesicles contained in cell culture supernatants and body fluid samples by flow cytometry. After extracellular vesicles are reacted and immobilized on magnetic beads on which extracellular vesicle surface phosphatidylserine (PS) specifically binding protein is immobilized, fluorescent labeling for any extracellular vesicle marker protein by using antibodies, we can detect marker proteins on extracellular vesicle surface with high sensitivity. Before using it, prepare the primary antibody for each surface marker protein, the fluorescent dye-labeled secondary antibody, or the primary antibody labeled with fluorescent dye.