Learn More
FUJIFILM Biosciences PS Capture™ Exosome Isolation Resin Kit, 1Kit(0.5 mL Slurry)
Catalog No. NC1700319
Encompass_Preferred
PS Capture™ Exosome Isolation Resin Kit. Kit Compentents: Exosome Capture Resin 0.5 mL x 1, Exosome Basic Buffer(10 x) 50 mL x 1, Binding Enhancer(100 x) 10 mL x 1, Washing Enhancer(100 x) 5 mL x 1, Eluting Enhancer(100 x) 0.5 mL x 1.
Catalog No. NC1700319 Supplier FUJIFILM Biosciences Supplier No. 29080301
PS Capture™ Exosome Isolation Resin Kit. Kit Compentents: Exosome Capture Resin 0.5 mL x 1, Exosome Basic Buffer(10 x) 50 mL x 1, Binding Enhancer(100 x) 10 mL x 1, Washing Enhancer(100 x) 5 mL x 1, Eluting Enhancer(100 x) 0.5 mL x 1.