Learn More
FUJIFILM BIOSCIENCES INC Exosome ELISA Kit (Anti Mouse IgG POD)
Supplier:  FUJIFILM BIOSCIENCES INC 29779201
Encompass_Preferred
This kit is an enzyme immunoassay reagent that can be used for the qualitative analysis of extracellular vesicles purified from cell culture supernatants and body fluid specimens and quantitative analysis of extracellular vesicles in cell culture supernatant specimens. Extracellular vesicles with optional marker protein on the surface can be detected with high sensitivity by using a solid phase immobilized protein that binds specifically to phosphatidylserine (PS) of extracellular vesicles and an arbitrary detection mouse monoclonal primary antibody.
Catalog No. NC1543956