Learn More
ILEX LIFE SCIENCES LLC PS CC DISH 100X15MM STRL 375CT
SDP
Supplier:  ILEX LIFE SCIENCES LLC CELL1000
Encompass_Preferred
Planet-Safe Cell Culture Dish, PLA-Based, Clear, Sterile, 100 mm x 15 mm, 375/case. 100% Plant-Based Planet-Safe Sustainable Labware is made from polylactic acid (PLA), a plant-based resin derived from sustainably cultivated sugarcane. USDA BioPreferred label. CELL GROWTH: Increased hydrophilic properties of Planet-Safe PLA surfaces allow a variety of anchorage-dependent cell species to grow on the dish surface without added coatings or treatments. IMPROVED CELL ADHESION: Planet-Safe surfaces exhibit improved cellular adhesion when compared to a leading brand. RELIABLE CELL GROWTH: identical growth rates were observed between Planet-Safe and Corning surfaces. Observed growth rate and confluence results were comparable to leading TC-treated polystyrene dishes. INCREASED STABILITY AND REPRODUCIBILITY: exhibits a much lower percentage of fluid shear compared to other available popular brands. COMPOSTABLE: commercially compostable per ASTM D6400 standards.
Catalog No. 50-273-3953