Learn More
Electron Microscopy Sciences POLYSTYRENE LATEX SIZE 0.204 µm
SDP
Catalog No. 5034506
Encompass_Preferred
Uniform polystyrene latex (size 0.204 microns) is suspended on a Formvar/Carbon grid and is usefule for checking magnification and calibration.
Catalog No. 50-345-06 Supplier Electron Microscopy Sciences Supplier No. 80030
Uniform polystyrene latex (size 0.204 microns) is suspended on a Formvar/Carbon grid and is usefule for checking magnification and calibration.