Learn More
Echelon Biosciences Research Labs PS LIPID MICROPARTICLES 100UL
SDP
Catalog No. NC1972401
Encompass
PS Lipid Microparticles contain attached phosphatidylserine with the phospholipid headgroups exposed and available for biological interactions. Phosphatidylserine (PS) is an anionic, intracellular phospholipid component of the cell membrane and is involved in cell signaling including apoptosis. This item is non returnable
This item is not returnable. View return policy
Catalog No. NC1972401 Supplier Echelon Biosciences Research Labs Supplier No. PB1PS100UL
PS Lipid Microparticles contain attached phosphatidylserine with the phospholipid headgroups exposed and available for biological interactions. Phosphatidylserine (PS) is an anionic, intracellular phospholipid component of the cell membrane and is involved in cell signaling including apoptosis. This item is non returnable