Learn More
Medchemexpress LLC HY-121744 5mg Medchemexpress, PS10 CAS:1564265-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1217445MG
Encompass_Preferred
Medchemexpress, HY-121744 5mg PS10 CAS:1564265-82-2 PS10 is a novel, potent and ATP-competitive pan-PDK inhibitor, inhibits all PDK isoforms with IC50 of 0.8 μM, 0.76 μM, 2.1 μM and 21.3 μM for PDK2, PDK4, PDK1, and PDK3, respectively. PS10 shows high affinity for PDK2 (Kd= 239 nM) than for Hsp90 (Kd= 47 μM). PS10 improves glucose tolerance, stimulates myocardial carbohydrate oxidation in diet-induced obesity. PS10 has the potential for the investigation of diabetic cardiomyopathy[2].PDK: pyruvate dehydrogenase kinase Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9913