Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7031MSM1XXP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM313. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Spent medium of MCF-7 cells, treated with estrogen. Normal stomach. About 60% of breast carcinomas are positive for pS2, especially those that are also positive for estrogen,/or progesterone receptor. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 6.5kDa.Chromosome: 16p11.2. Gene Symbol: TFF1. Gene ID: 7031. Ensemble: ENSG00000160182. Uniprot: NP_003216.
Catalog No. 50-188-5200