Learn More
Medchemexpress LLC PS372424 hydrochloride | 1596362-29-6 | ≥98.0% | 625.20 | 10 MG
SDP
Catalog No. 5000343201
Encompass_Preferred
PS372424 hydrochloride, a three amino-acid fragment of CXCL10, is a specific human CXCR3 agonist with anti-inflammatory activity. It prevents human T-cell migration in a humanized model of arthritic inflammation.
  • Specific human CXCR3 agonist
  • Anti-inflammatory activity
  • Prevents human T-cell migration in a humanized model of arthritic inflammation
  • For research use only
Catalog No. 50-003-43201 Supplier Medchemexpress LLC Supplier No. HY111149A10MM/1ML
PS372424 hydrochloride, a three amino-acid fragment of CXCL10, is a specific human CXCR3 agonist with anti-inflammatory activity. It prevents human T-cell migration in a humanized model of arthritic inflammation.
  • Specific human CXCR3 agonist
  • Anti-inflammatory activity
  • Prevents human T-cell migration in a humanized model of arthritic inflammation
  • For research use only