Learn More
Medchemexpress LLC HY-13851 5mg Medchemexpress, PS47 CAS:1180676-33-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY138515MG
Encompass_Preferred
Medchemexpress, HY-13851 5mg PS47 CAS:1180676-33-8 PS47 is an inactive E-isomer of PS48. PS48 is an activator of PDK1. PS47 can be used as a negative control for PS48. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1525