Learn More
Electron Microscopy Sciences PS4R with Grats certificate
SDP
Catalog No. 501901568
Encompass_Preferred
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions
Catalog No. 50-190-1568 Supplier Electron Microscopy Sciences Supplier No. 68047PS4RGR
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions