Learn More
Electron Microscopy Sciences PS4R with NPL Certificate
SDP
Catalog No. 501901570
Encompass_Preferred
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions
Catalog No. 50-190-1570 Supplier Electron Microscopy Sciences Supplier No. 68047PS4RNP
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions