Learn More
Electron Microscopy Sciences USAF Test Chart PS75N Negative Image
SDP
Catalog No. 501901577
Encompass_Preferred
Metal foil version of the USAF test chart has clear, negative, high contrast patterns in groups -2 to 4. Absence of a glass support substrate removes the problems of masking infrared regions.
Type: Negative Image
Use: Groups 2/1 to 9/3
Catalog No. 50-190-1577 Supplier Electron Microscopy Sciences Supplier No. 6809806
Metal foil version of the USAF test chart has clear, negative, high contrast patterns in groups -2 to 4. Absence of a glass support substrate removes the problems of masking infrared regions.
Type: Negative Image
Use: Groups 2/1 to 9/3