Learn More
Genscript Corporation PSA 7H2 mAb Mouse
Catalog No. 5000058788
Encompass
Prostate specific antigen (PSA) is a serine protease produced by the prostate gland In serum it exists in different forms cPSA mainly including PSA-ACT (alpha-1-antichymotrypsin) and PSA-A2M (alpha-2-macroglobulin) fPSA (free PSA) etc The blood level of PSA is increased in men with prostate cancer and it is considered as an indicator for prostate cancer tumor recurrence and response to therapy
Catalog No. 50-000-58788 Supplier Genscript Corporation Supplier No. V07302100
Add to Cart
Add to Cart
Request a Quote
Prostate specific antigen (PSA) is a serine protease produced by the prostate gland In serum it exists in different forms cPSA mainly including PSA-ACT (alpha-1-antichymotrypsin) and PSA-A2M (alpha-2-macroglobulin) fPSA (free PSA) etc The blood level of PSA is increased in men with prostate cancer and it is considered as an indicator for prostate cancer tumor recurrence and response to therapy