Learn More
Genscript Corporation PSA 7H2 mAb Mouse
Catalog No. 5000057039
Encompass
Prostate specific antigen (PSA) is a serine protease produced by the prostate gland In serum it exists in different forms cPSA mainly including PSA-ACT (alpha-1-antichymotrypsin) and PSA-A2M (alpha-2-macroglobulin) fPSA (free PSA) etc The blood level of PSA is increased in men with prostate cancer and it is considered as an indicator for prostate cancer tumor recurrence and response to therapy
Catalog No. 50-000-57039 Supplier Genscript Corporation Supplier No. V073021
Prostate specific antigen (PSA) is a serine protease produced by the prostate gland In serum it exists in different forms cPSA mainly including PSA-ACT (alpha-1-antichymotrypsin) and PSA-A2M (alpha-2-macroglobulin) fPSA (free PSA) etc The blood level of PSA is increased in men with prostate cancer and it is considered as an indicator for prostate cancer tumor recurrence and response to therapy