Learn More
Drg International Inc Psa Free Elisa Ruo
Catalog No. 501947607
Encompass_Preferred
An enzyme immunoassay for the quantitative determination of free prostate specific antigen (f-PSA) in serum or plasma. This product is for Research Use Only. This f-PSA ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA) . The microtiter wells are coated with an anti-f-PSA monoclonal antibody, directed towards an epitope of an antigen molecule.
Catalog No. 50-194-7607 Supplier Drg International Inc Supplier No. EIA4189R
An enzyme immunoassay for the quantitative determination of free prostate specific antigen (f-PSA) in serum or plasma. This product is for Research Use Only. This f-PSA ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA) . The microtiter wells are coated with an anti-f-PSA monoclonal antibody, directed towards an epitope of an antigen molecule.