Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PSA (Free)/KLK3 ELISA Kit

Catalog No. EHKLK3FX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHKLK3FX5 5 x 96 Tests
EHKLK3F 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHKLK3FX5 Supplier Invitrogen™ Supplier No. EHKLK3FX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Human Free Prostate Specific Antigen/Kallikrein 3 (Hu freePSA/KLK3) ELISA quantitates Hu free PSA/KLK3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu free PSA/KLK3. Principle of the method The Human free PSA/KLK3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and is abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07288
Assay Range 45-12000 pg/mL
Assay Sensitivity 45 pg/mL
Calibration Range 45 pg/mL to 12,000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF-7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF,SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Antigen; antigen, prostate specific; APS; Fletcher factor; gamma-seminoprotein; hK3; Kal3; Kal-3; Kallikrein 3; kallikrein 3, plasma; kallikrein B, plasma 1; kallikrein related peptidase 3; kallikrein-3; Kallikrein-3 (KLK3); kallikrein-related peptidase 3; Kalp1; KALP15; Kininogenin; KLK2A1; Klk3; KLK-3; Klkb1; P-30 antigen; Pk; Plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; prostate specific antigen; prostate-specific (APS); prostate-specific antigen; PSA; Semenogelase; semininAntigen; antigen, prostate specific; APS; Fletcher factor; gamma-seminoprotein; hK3; Kal3; Kal-3; Kallikrein 3; kallikrein 3, plasma; kallikrein B, plasma 1; kallikrein related peptidase 3; kallikrein-3; Kallikrein-3 (KLK3); kallikrein-related peptidase 3; Kalp1; KALP15; Kininogenin; KLK2A1; Klk3; KLK-3; Klkb1; P-30 antigen; Pk; Plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; prostate specific antigen; prostate-specific (APS); prostate-specific antigen; PSA; Semenogelase; seminin;
Gene ID (Entrez) 354
Gene Symbol KLK3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases PSA, Gamma-seminoprotein, Seminin, Kallikrein-3, P-30 antigen, Semenogelase
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Angiogenesis
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym APS, KLK2A1, PSA, hK3
Target PSA
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.