Learn More
Gentarget PSA-GFP PURO LENTIVIRUS
Catalog No. NC2586123
Encompass
Pre-made lentivirus express GFP fluorescent reporter under a native human PSA promoter for selective expression in Prostate tissue or Cells.This lentivirus also contain the Puromycin selection marker under the constitutive RSV promoter for antibiotic selection. Amount: 200ul/per vial, at 1 x 107 IFU/ml in DMEM medium with 10% FBS.
Catalog No. NC2586123 Supplier Gentarget Supplier No. LVP1045P
Pre-made lentivirus express GFP fluorescent reporter under a native human PSA promoter for selective expression in Prostate tissue or Cells.This lentivirus also contain the Puromycin selection marker under the constitutive RSV promoter for antibiotic selection. Amount: 200ul/per vial, at 1 x 107 IFU/ml in DMEM medium with 10% FBS.