Learn More
Cayman Chemical PSA RabbIt MoncLNal ntIbod
SDP
Catalog No. 502948565
Encompass_Preferred
A mixture of ganglioside GM3 with variable fatty acyl chains demonstrates antiproliferative and pro-apoptotic effects in tumor cells by modulating cell adhesion proliferation and differentiation suppresses angiogenesis and reduces proliferation and migration of HUVECs at 20 M induces dissociation of the insulin receptor-caveolin-1 complex from lipid microdomains
Catalog No. 50-294-8565 Supplier Cayman Chemical Supplier No. 32273100 UL
A mixture of ganglioside GM3 with variable fatty acyl chains demonstrates antiproliferative and pro-apoptotic effects in tumor cells by modulating cell adhesion proliferation and differentiation suppresses angiogenesis and reduces proliferation and migration of HUVECs at 20 M induces dissociation of the insulin receptor-caveolin-1 complex from lipid microdomains