Learn More
Carbosynth LLC. Psalmotoxin 1 (0.1mg vial), 1ea, Disulfide bonds: (Disulfide bonds between Cys3-Cys18, Cys10-Cys23, and Cys17-Cys33), MW: 4689.4, Molecular formula: C200H312N62O57S6
SDP
Supplier:  Carbosynth LLC. PTX4435S0.1MG
Encompass_Preferred
Synonyms: Glu-Asp-Cys-Ile-Pro-Lys-Trp-Lys-Gly-Cys-Val-Asn-Arg-His-Gly-Asp-Cys-Cys-Glu-Gly-Leu-Glu-Cys-Trp-Lys-Arg-Arg-Arg-Ser-Phe-Glu-Val-Cys-Val-Pro-Lys-Thr-Pro-Lys-Thr, Application: Selective Blocker for Acid-Sensitive Ion Channel, ASIC1a, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: P. Escoubas, J.R. De Weille, A. Lecoq, S. Diochot, R. Waldmann, G. Champigny, D. Moinier, A. Menez, and M. Lazdunski, J. Biol. Chem., 275, 25116 (2000). (Original; Primary Structure & ASIC Blocking Selectivity), P. Escoubas, C. Bernard, G. Lambeau, M. Lazdunski, and H. Darbon, Protein Sci., 12, 1332 (2003). (Three-dimensional Solution Structure), X. Chen, H. Kalbacher, S. Grunder, J. Gen. Physiol., 127, 267 (2006). (Pharmacol.; State-Dependent Function)
Catalog No. 50-168-6805