Learn More
Medchemexpress LLC Psalmotoxin 1 TFA | 880107-52-8 | 99.37% | 4689.39 | 500 UG
SDP
Catalog No. 5000422975
Encompass_Preferred
Psalmotoxin 1 is a protein toxin that binds at subunit-subunit interfaces of acid-sensing ion channel 1a (ASIC1a). It acts as a potent and selective ASIC1a inhibitor by increasing the apparent affinity for H+ of ASIC1a. Psalmotoxin 1 can induce cell apoptosis and inhibit cell migration, proliferation, and invasion of cancer cells. It is utilized in research concerning cancers and neurological diseases.
  • Potent and selective ASIC1a inhibitor with an IC50 of 0.9 nM
  • Induces cell apoptosis
  • Inhibits cell migration, proliferation, and invasion of cancer cells
  • Used in research concerning cancers and neurological diseases
  • Neuroprotective in a conscious model of stroke
  • Inhibits tumor growth in breast cancer models
Catalog No. 50-004-22975 Supplier Medchemexpress LLC Supplier No. HYP1411A500UG
Psalmotoxin 1 is a protein toxin that binds at subunit-subunit interfaces of acid-sensing ion channel 1a (ASIC1a). It acts as a potent and selective ASIC1a inhibitor by increasing the apparent affinity for H+ of ASIC1a. Psalmotoxin 1 can induce cell apoptosis and inhibit cell migration, proliferation, and invasion of cancer cells. It is utilized in research concerning cancers and neurological diseases.
  • Potent and selective ASIC1a inhibitor with an IC50 of 0.9 nM
  • Induces cell apoptosis
  • Inhibits cell migration, proliferation, and invasion of cancer cells
  • Used in research concerning cancers and neurological diseases
  • Neuroprotective in a conscious model of stroke
  • Inhibits tumor growth in breast cancer models