Learn More
AdipoGen Psammaplin A
SDP
Supplier:  AdipoGen AGCN20088C100
Encompass_Preferred
Chemical. CAS 110659-91-1. Formula C22H24Br2N4O6S2. MW 664.4. Isolated from Psammaplysilla purpurea. Antibiotic. Caffeine-like Ca2 releaser in heavy sarcoplasmic reticulum HSR. Antifungal. Chitinase inhibitor. DNA methyltransferase inhibitor. Histone deacetylase HDAC inhibitor. Anticancer compound. Angiogenesis suppressor.
Catalog No. 50-256-2665