Learn More
ABclonal Technology PSAP
SDP
Catalog No. 5000007803
Encompass_Preferred
This gene encodes an enzyme that catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland An alternatively spliced transcript variant encoding a longer isoform has been found for this gene This isoform contains a transmembrane domain and is localized in the plasma membrane-endosomal-lysosomal pathway
Catalog No. 50-000-07803 Supplier ABclonal Technology Supplier No. A2119520UL
This gene encodes an enzyme that catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland An alternatively spliced transcript variant encoding a longer isoform has been found for this gene This isoform contains a transmembrane domain and is localized in the plasma membrane-endosomal-lysosomal pathway