Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 55MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: ACPP/1338. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: PC12 cells or normal prostate or prostate carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length ACPP protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 52kDa.Chromosome: 3q22.1. Gene Symbol: ACPP. Gene ID: 55. Ensemble: ENSG00000014257. Uniprot: NP_001127666.
Catalog No. 50-188-5079