Learn More
Medchemexpress LLC Psar18-cooh | 2313534-26-6 | 96.0% | 1339.45 g/mol | C56H94N18O20 | 10 MG
SDP
Catalog No. 5000285351
Encompass_Preferred
PSAR18-COOH is a carboxyl-terminated polysarcosine (PSAR) derivative for research use. It provides a hydrophilic, biodegradable, non-immunogenic, and water-soluble polymer backbone suitable for use in drug and diagnostic delivery system development.
  • Hydrophilic and water-soluble polymer suitable for biological delivery systems.
  • Biodegradable and non-immunogenic, reducing potential immune response.
  • Carboxyl terminal group enables conjugation or further chemical modification.
  • High purity suitable for research applications.
  • Defined molecular characteristics for reproducible experimental use.
  • Recommended storage conditions to preserve stability: cold, sealed, and protected from moisture and light.
Catalog No. 50-002-85351 Supplier Medchemexpress LLC Supplier No. HY16042910MG
Add to Cart
Add to Cart
PSAR18-COOH is a carboxyl-terminated polysarcosine (PSAR) derivative for research use. It provides a hydrophilic, biodegradable, non-immunogenic, and water-soluble polymer backbone suitable for use in drug and diagnostic delivery system development.
  • Hydrophilic and water-soluble polymer suitable for biological delivery systems.
  • Biodegradable and non-immunogenic, reducing potential immune response.
  • Carboxyl terminal group enables conjugation or further chemical modification.
  • High purity suitable for research applications.
  • Defined molecular characteristics for reproducible experimental use.
  • Recommended storage conditions to preserve stability: cold, sealed, and protected from moisture and light.