Learn More
Medchemexpress LLC PSAR18-COOH | 2313534-26-6 | 99.5% | 1339.45 | 25 MG
SDP
Catalog No. 5000392582
Encompass_Preferred
PSAR18-COOH is a derivative of PSAR, which is extracted from patent WO2009064913A1. PSAR is a highly hydrophilic, biodegradable, non-immunogenic, and water-soluble polymer used in various delivery systems for drugs or diagnostics.
  • Derivative of PSAR.
  • Highly hydrophilic.
  • Biodegradable.
  • Non-immunogenic.
  • Water-soluble polymer.
  • Employed in several delivery systems for drugs or diagnostics.
Catalog No. 50-003-92582 Supplier Medchemexpress LLC Supplier No. 25MGHY160429
Add to Cart
Add to Cart
PSAR18-COOH is a derivative of PSAR, which is extracted from patent WO2009064913A1. PSAR is a highly hydrophilic, biodegradable, non-immunogenic, and water-soluble polymer used in various delivery systems for drugs or diagnostics.
  • Derivative of PSAR.
  • Highly hydrophilic.
  • Biodegradable.
  • Non-immunogenic.
  • Water-soluble polymer.
  • Employed in several delivery systems for drugs or diagnostics.