Learn More
Medchemexpress LLC PSB-0739 | 1052087-90-7 | 98.0% | 609.54 | C26H17N3Na2O8S2 | 5 MG
SDP
Catalog No. 5000263288
Encompass_Preferred
PSB-0739 is a research-grade P2Y12 receptor antagonist used for laboratory studies of platelet aggregation and P2Y12 signaling. It is a high-affinity, potent, competitive, non-nucleotide small molecule (CAS 1052087-90-7; MW 609.54) supplied as a solid intended for research use only.
  • High-affinity P2Y12 receptor antagonist (Ki = 24.9 nM).
  • Potent competitive non-nucleotide antagonist in vitro (pA2 = 9.8).
  • Inhibits ADP-evoked Ca2+ responses (EC50 ≈ 5.4 μM).
  • Purity 98.0%.
  • Intended for research use only.
  • Store sealed at 4°C; in solvent: -80°C for long term.
Catalog No. 50-002-63288 Supplier Medchemexpress LLC Supplier No. HY1086605MG
PSB-0739 is a research-grade P2Y12 receptor antagonist used for laboratory studies of platelet aggregation and P2Y12 signaling. It is a high-affinity, potent, competitive, non-nucleotide small molecule (CAS 1052087-90-7; MW 609.54) supplied as a solid intended for research use only.
  • High-affinity P2Y12 receptor antagonist (Ki = 24.9 nM).
  • Potent competitive non-nucleotide antagonist in vitro (pA2 = 9.8).
  • Inhibits ADP-evoked Ca2+ responses (EC50 ≈ 5.4 μM).
  • Purity 98.0%.
  • Intended for research use only.
  • Store sealed at 4°C; in solvent: -80°C for long term.